4-Ethynylbenzaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: p-Ethynylbenzaldehyde, 4-ethynyl-benzaldehyde, Benzaldehyde,4-ethynyl-

A234214-1G A234214-5G A234214-10G
103810-930EA 6 USD
103810-930 103850-690 77089-666 77089-668 77568-276
4-Ethynylbenzaldehyde 98%
4-Ethynylbenzaldehyde

Precaución: For research use only.

Formula: C₉H₆O
Boiling Pt: 223.8 °C (760 mm Hg)
Melting Pt: 89…93 °C
Flash Pt: 86.8 °C
Storage Temperature: Freezer
MDL Number: MFCD05664348
CAS Number: 63697-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR