4-Ethynylanisole 98% (by GC)

Supplier: Ambeed
Warning

A154389-100G A154389-10G A154389-500G A154389-1G A154389-25G A154389-5G
103804-636EA 9 USD
103804-636 103842-824 77259-404 77259-406 77259-408 77300-056 77318-542 77318-544
4-Ethynylanisole 98% (by GC)
4-Ethynylanisole

Precaución: for Research Use Only

Formula: C₉H₈O
Boiling Pt: 194.8 °C (760 mm Hg)
Melting Pt: 28…29 °C
Flash Pt: 82 °C
Storage Temperature: Ambient
MDL Number: MFCD00168815
CAS Number: 768-60-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR