4-Ethynylaniline 98%

Supplier: Ambeed
Warning

Synonyms: (4-Aminophenyl)acetylene, 1-Ethynyl-4-aminobenzene, 1-Amino-4-ethynylbenzene, (4-Aminophenyl)ethyne

A168204-1G A168204-25G A168204-5G A168204-100G A168204-10G A168204-250MG
103806-264EA 6 USD
103806-264 103844-522 77081-574 77081-576 77081-578 77236-190 77236-192 77236-194
4-Ethynylaniline 98%
4-Ethynylaniline

Precaución: for Research Use Only

Formula: C₈H₇N
MW: 117.15 g/mol
Melting Pt: 98…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00168824
CAS Number: 14235-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR