4-Ethyl-5-fluoropyrimidine 97%

Supplier: Ambeed
Warning

A397659-1G A397659-25G A397659-5G A397659-10G
103827-002EA 15 USD
103827-002 103838-042 76841-472 76841-474 76841-476 77115-704
4-Ethyl-5-fluoropyrimidine 97%
4-Ethyl-5-fluoropyrimidine

Precaución: for Research Use Only

Formula: C₆H₇FN₂
Boiling Pt: 169.5 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06657566
CAS Number: 137234-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR