4-Ethylbiphenyl 95%

Supplier: Ambeed
Warning

Synonyms: 4-Ethyl-1,1'-Biphenyl

A628923-1G A628923-25G A628923-5G A628923-10G
103840-268EA 12 USD
103840-268 76844-430 76844-432 76844-434 77035-332
4-Ethylbiphenyl 95%
4-Ethylbiphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₄
MW: 182.27 g/mol
Boiling Pt: 287.9±10.0 °C (760 mm Hg)
Melting Pt: 34…35.5 °C
Flash Pt: 128.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00010236
CAS Number: 5707-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR