4-Cyano-4'-ethylbiphenyl 98%

Supplier: Ambeed
Danger

Synonyms: 4'-Ethyl-[1,1'-biphenyl]-4-carbonitrile

A529174-25G A529174-5G
103839-666EA 14 USD
103839-666 77307-728 77307-730
4-Cyano-4'-ethylbiphenyl 98%
4-Cyano-4'-ethylbiphenyl

Precaución: for Research Use Only

Formula: C₁₅H₁₃N
MW: 207.27 g/mol
Boiling Pt: 354.0±31.0 °C (760 mm Hg)
Melting Pt: 74…76 °C
Flash Pt: 168.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00799420
CAS Number: 58743-75-2
UN: 3439
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR