4'-Ethoxyacetophenone 95%

Supplier: Ambeed
Warning

A868295-100G A868295-1G A868295-25G A868295-5G
103841-272EA 14 USD
103841-272 77036-740 77308-464 77308-466 77308-468
4'-Ethoxyacetophenone 95%
4'-Ethoxyacetophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 268…269 °C at 758 mmHg
Melting Pt: 35…38 °C at N/A mmHg
Flash Pt: 134 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00009095
CAS Number: 1676-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR