4-Ethoxy-2-fluoropyridine 97%

Supplier: Ambeed
Warning

A158902-100MG A158902-250MG A158902-1G
103805-188EA 175 USD
103805-188 103843-400 77286-380 77286-382 77300-376
4-Ethoxy-2-fluoropyridine 97%
4-Ethoxy-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₇H₈FNO
Boiling Pt: 187.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09835274
CAS Number: 175965-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR