4-Dodecylaniline 97%

Supplier: Ambeed
Warning

Synonyms: 4-Dodecylphenylamine, 1-Amino-4-dodecylbenzene

A200471-10G A200471-1G A200471-5G
103817-204EA 11 USD
103817-204 103847-832 77030-110 77302-682 77302-684
4-Dodecylaniline 97%
4-Dodecylaniline

Precaución: for Research Use Only

Formula: C₁₈H₃₁N
MW: 261.45 g/mol
Boiling Pt: 220…221 °C (760 mm Hg)
Melting Pt: 35…39 °C
Flash Pt: 150 °C
Storage Temperature: Ambient
MDL Number: MFCD00007919
CAS Number: 104-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR