4-Dimethoxymethylpyrimidin-2-ylamine 97%

Supplier: Ambeed
Warning

A111199-100G A111199-10G A111199-250MG A111199-25G A111199-1G A111199-5G
103797-800EA 11 USD
103797-800 103823-612 77008-928 77008-930 77008-932 77008-934 77310-418 77310-420
4-Dimethoxymethylpyrimidin-2-ylamine 97%
4-Dimethoxymethylpyrimidin-2-ylamine

Precaución: for Research Use Only

Formula: C₇H₁₁N₃O₂
MW: 169.1800 g/mol
Boiling Pt: 295.3 °C (760 mm Hg)
Melting Pt: 158…162 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05864792
CAS Number: 165807-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR