Dibenzothiophene-4-boronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Dibenzothiopheneboronic acid
A119739-100G
A119739-10G
A119739-25G
A119739-1G
A119739-5G
103799-826EA
5
USD
103799-826
77284-768
77284-770
77284-772
77297-716
77311-928
Dibenzothiophene-4-boronic acid 98%
Dibenzothiophene-4-boronic acid
Precaución: for Research Use Only
Formula:
C₁₂H₉BO₂S MW: 228.08 g/mol Boiling Pt: 480.2 °C (760 mm Hg) Melting Pt: 327…330 °C Flash Pt: 244.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318182 CAS Number: 108847-20-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...