4-Cyclopropylnaphthalen-1-amine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A113828-10G
A113828-250MG
A113828-1G
A113828-5G
103798-300EA
6
USD
103798-300
103824-142
77189-200
77189-202
77297-270
77297-272
4-Cyclopropylnaphthalen-1-amine hydrochloride 97%
4-Cyclopropylnaphthalen-1-amine hydrochloride
Precaución: for Research Use Only
Formula:
C₁₃H₁₄ClN MW: 219.71 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28387394 CAS Number: 1533519-92-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...