4-Cyclopropylbenzaldehyde 96%

Supplier: Ambeed
Warning

A127070-100MG A127070-250MG
103800-908EA 102 USD
103800-908 77026-872 77599-846
4-Cyclopropylbenzaldehyde 96%
4-Cyclopropylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀O
Storage Temperature: Refrigerator
MDL Number: MFCD06802398
CAS Number: 20034-50-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR