4-Cyclohexylaniline 97%

Supplier: Ambeed
Warning

A173712-5G A173712-100MG A173712-10G A173712-250MG A173712-1G
103806-862EA 9 USD
103806-862 103845-162 77082-432 77286-948 77286-950 77286-952 77301-294
4-Cyclohexylaniline 97%
4-Cyclohexylaniline

Precaución: for Research Use Only

Formula: C₁₂H₁₇N
MW: 175.27 g/mol
Boiling Pt: 301.2 °C (760 mm Hg)
Melting Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00001454
CAS Number: 6373-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR