4-Cyano-4-{[(ethylsulfanyl)carbonothioyl]sulfanyl}pentanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-Cyano-4-(((ethylthio)carbonothioyl)thio)pentanoic acid

A718802-100MG A718802-250MG A718802-5G
103840-724EA 944 USD
103840-724 77255-308 77255-310 77308-188
4-Cyano-4-{[(ethylsulfanyl)carbonothioyl]sulfanyl}pentanoic acid 95%
4-Cyano-4-{[(ethylsulfanyl)carbonothioyl]sulfanyl}pentanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₃NO₂S₃
MW: 263.4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27978407
CAS Number: 1137725-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR