4-Cyano-3-fluorophenyl 4-(trans-4-pentylcyclohexyl)benzoate 97%

Supplier: Ambeed
Warning

A100033-10G A100033-250MG A100033-25G
103795-214EA 7 USD
103795-214 103820-936 77270-132 77270-134 77270-136
4-Cyano-3-fluorophenyl 4-(trans-4-pentylcyclohexyl)benzoate 97%
4-Cyano-3-fluorophenyl 4-(trans-4-pentylcyclohexyl)benzoate

Precaución: for Research Use Only

Formula: C₂₅H₂₈FNO₂
MW: 393.49 g/mol
Boiling Pt: 527.0±50.0 °C (760 mm Hg)
Flash Pt: 272.5 °C
Storage Temperature: Ambient
MDL Number: MFCD11053367
CAS Number: 92118-84-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR