4-Cyano-1H-indole-6-carboxylic acid 95%

Supplier: Ambeed
Warning

A264462-100MG A264462-1G A264462-250MG
103792-520EA 25 USD
103792-520 103812-680 77098-884 77098-886 77098-888
4-Cyano-1H-indole-6-carboxylic acid 95%
4-Cyano-1H-indole-6-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆N₂O₂
Boiling Pt: 484.8±30.0 °C (760 mm Hg)
Flash Pt: 247 °C
Storage Temperature: Refrigerator
MDL Number: MFCD23704066
CAS Number: 1427502-44-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR