Chroman-4-one 97%

Supplier: Ambeed
Warning

A113046-100G A113046-10G A113046-1G A113046-25G A113046-500G A113046-5G
103823-992EA 7 USD
103823-992 77284-268 77284-270 77284-272 77284-274 77284-276 77297-224
Chroman-4-one 97%
Chroman-4-one

Precaución: for Research Use Only

Formula: C₉H₈O₂
MW: 148.16 g/mol
Boiling Pt: 127…128 °C at 13 mmHg
Melting Pt: 35…38 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00006840
CAS Number: 491-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR