4-Chlorothieno[2,3-d]pyrimidine 98%

Supplier: Ambeed
Warning

A124628-10G A124628-250MG A124628-25G A124628-5G
103800-558EA 23 USD
103800-558 77149-362 77149-364 77149-366 77298-010
4-Chlorothieno[2,3-d]pyrimidine 98%
4-Chlorothieno[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₃ClN₂S
MW: 170.6200 g/mol
Boiling Pt: 285.7 °C (760 mm Hg)
Melting Pt: 109…113 °C
Flash Pt: 126.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01312373
CAS Number: 14080-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR