4-Chlorothiazole-5-carboxaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 4-Chlorothiazole-5-carbaldehyde

A320857-1G A320857-5G A320857-10G A320857-250MG A320857-25G
103825-254EA 55 USD
103825-254 103836-186 76839-070 76839-072 77290-290 77290-292 77306-228
4-Chlorothiazole-5-carboxaldehyde 97%
4-Chlorothiazole-5-carboxaldehyde

Precaución: for Research Use Only

Formula: C₄H₂ClNOS
MW: 147.5828 g/mol
Boiling Pt: 254.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09837291
CAS Number: 104146-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR