4-Chloroquinoline-3-carbonitrile 97%

Supplier: Ambeed
Warning

A269840-250MG A269840-1G
103812-980EA 112 USD
103812-980 103852-886 77289-406 77305-038
4-Chloroquinoline-3-carbonitrile 97%
4-Chloroquinoline-3-carbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₅ClN₂
Boiling Pt: 348.234 °C (760 mm Hg)
Flash Pt: 164.406 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03093327
CAS Number: 69875-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR