4-Chloroquinoline 97%
Supplier: Ambeed
|
Warning
|
A158683-100G
A158683-10G
A158683-250MG
A158683-500G
A158683-1G
A158683-25G
A158683-5G
103805-158EA
9
USD
103805-158
103843-372
77110-564
77110-566
77110-568
77110-570
77300-352
77300-354
77300-356
4-Chloroquinoline 97%
4-Chloroquinoline
Precaución: for Research Use Only
Formula:
C₉H₆ClN MW: 163.61 g/mol Boiling Pt: 255.8 °C (760 mm Hg) Melting Pt: 28…31 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006773 CAS Number: 611-35-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...