4-Chloroquinolin-8-amine 95%

Supplier: Ambeed
Warning

A170087-250MG A170087-1G
103806-448EA 37 USD
103806-448 103844-714 76824-464 77081-812
4-Chloroquinolin-8-amine 95%
4-Chloroquinolin-8-amine

Precaución: for Research Use Only

Formula: C₉H₇ClN₂
Storage Temperature: Refrigerator
MDL Number: MFCD18448245
CAS Number: 81764-16-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR