4-Chloroquinolin-2(1H)-one 96%

Supplier: Ambeed
Warning

A401134-1G A401134-5G A401134-100MG A401134-250MG
103827-050EA 106 USD
103827-050 103838-086 76841-544 76841-546 76979-060 76979-062
4-Chloroquinolin-2(1H)-one 96%
4-Chloroquinolin-2(1H)-one

Precaución: For research use only

Formula: C₉H₆ClNO
Boiling Pt: 366.9 °C (760 mm Hg)
Flash Pt: 175.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00234492
CAS Number: 20146-59-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR