4-Chloroquinazoline 97%

Supplier: Ambeed
Danger

A587542-10G A587542-1G A587542-25G A587542-5G
103828-902EA 9 USD
103828-902 103840-048 77103-866 77307-888 77307-890 77307-892
4-Chloroquinazoline 97%
4-Chloroquinazoline

Precaución: for Research Use Only

Formula: C₈H₅ClN₂
MW: 164.5900 g/mol
Boiling Pt: 281.0±13.0 °C (760 mm Hg)
Melting Pt: 96…100 °C
Flash Pt: 150.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00228682
CAS Number: 5190-68-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR