4-Chloropyrido[3,4-d]pyrimidine 95%

Supplier: Ambeed
Warning

A121307-250MG A121307-1G
103800-058EA 171 USD
103800-058 76718-544 77312-180
4-Chloropyrido[3,4-d]pyrimidine 95%
4-Chloropyrido[3,4-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₄ClN₃
MW: 165.5798 g/mol
Boiling Pt: 316.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09999184
CAS Number: 51752-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR