4-Chloropyridine-2-carbonitrile 98%

Supplier: Ambeed
Warning

A393113-1G A393113-25G A393113-5G A393113-100G A393113-10G A393113-500G
103826-916EA 10 USD
103826-916 103837-956 76841-356 76841-358 76841-360 77016-214 77016-216 77016-218
4-Chloropyridine-2-carbonitrile 98%
4-Chloropyridine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₃ClN₂
MW: 138.5500 g/mol
Boiling Pt: 308.2 °C (760 mm Hg)
Melting Pt: 82…86 °C
Flash Pt: 93.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06009833
CAS Number: 19235-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR