4-Chloro-2,6-pyridinedicarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-Chloropyridine-2,6-dicarboxylic acid

A265318-1G A265318-25G A265318-5G A265318-10G A265318-250MG
103812-724EA 9 USD
103812-724 103852-618 77092-296 77092-298 77092-300 77289-324 77289-326
4-Chloro-2,6-pyridinedicarboxylic acid 97%
4-Chloro-2,6-pyridinedicarboxylic acid

Precaución: for Research Use Only

Formula: C₇H₄ClNO₄
MW: 201.57 g/mol
Boiling Pt: 457.1 °C (760 mm Hg)
Melting Pt: 210 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04117894
CAS Number: 4722-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR