4-Chloropyridine-2,3-diamine 97%

Supplier: Ambeed
Warning

Synonyms: 2,3-Diamino-4-chloropyridine

A115518-250MG A115518-1G A115518-5G
103798-580EA 29 USD
103798-580 103824-438 76781-376 77311-256 77311-258
4-Chloropyridine-2,3-diamine 97%
4-Chloropyridine-2,3-diamine

Precaución: for Research Use Only

Formula: C₅H₆ClN₃
MW: 143.57 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13181777
CAS Number: 24484-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR