4-Chloropyridine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 4-Chloropyridinium chloride

A389710-25G A389710-5G A389710-100G A389710-10G A389710-500G
103810-260EA 207 USD
103810-260 103837-892 76841-260 76841-262 77033-538 77033-540 77033-542
4-Chloropyridine hydrochloride 97%
4-Chloropyridine hydrochloride

Precaución: for Research Use Only

Formula: C₅H₅Cl₂N
MW: 150.01 g/mol
Flash Pt: 198 °C
Storage Temperature: Ambient
MDL Number: MFCD00012829
CAS Number: 7379-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR