2-Amino-4-chloropyridine 98%

Supplier: Ambeed
Warning

A148740-100G A148740-10G A148740-500G A148740-1G A148740-25G A148740-5G
103803-964EA 5 USD
103803-964 103842-082 77010-656 77010-658 77010-660 77299-682 77317-548 77317-550
2-Amino-4-chloropyridine 98%
2-Amino-4-chloropyridine

Precaución: for Research Use Only

Formula: C₅H₅ClN₂
Boiling Pt: 240.7 °C (760 mm Hg)
Melting Pt: 126…127 °C
Flash Pt: 99.4 °C
Storage Temperature: Ambient
MDL Number: MFCD04113820
CAS Number: 19798-80-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR