4-Chloropicolinic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloropyridine-2-carboxylic acid

A234819-1000G A234819-100G A234819-10G A234819-500G A234819-25G A234819-5G
103850-736EA 6 USD
103850-736 76976-296 76976-298 76976-300 76976-302 77089-734 77089-736
4-Chloropicolinic acid 98%
4-Chloropicolinic acid

Precaución: for Research Use Only

Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Boiling Pt: 308 °C (760 mm Hg)
Melting Pt: 181 °C
Flash Pt: 140.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00191400
CAS Number: 5470-22-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR