4-Chlorophenylhydrazinium chloride 97%

Supplier: Ambeed
Warning

Synonyms: 4-Chlorophenylhydrazine hydrochloride, 1-(4-Chlorophenyl)hydrazine hydrochloride

A170342-100G A170342-10G A170342-500G A170342-25G
103809-026EA 175 USD
103809-026 76998-184 76998-186 76998-188 77081-860
4-Chlorophenylhydrazinium chloride 97%
4-Chlorophenylhydrazinium chloride

Precaución: for Research Use Only

Formula: C₆H₈Cl₂N₂
MW: 179.05 g/mol
Melting Pt: 216 °C
Storage Temperature: Ambient
MDL Number: MFCD00012943
CAS Number: 1073-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR