4-Chlorobenzoylacetonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 4-Chlorophenacylcyanide, 3-(4-Chlorophenyl)-3-oxopropanenitrile

A430835-100G A430835-10G A430835-25G A430835-5G A430835-1G
103827-514EA 6 USD
103827-514 103838-580 76695-054 76695-056 76842-272 76842-274 77307-250
4-Chlorobenzoylacetonitrile 97%
4-Chlorobenzoylacetonitrile

Precaución: for Research Use Only

Formula: C₉H₆ClNO
Melting Pt: 129…133 °C
Flash Pt: 165.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00051625
CAS Number: 4640-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR