4-Chloro-N-methylpicolinamide hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A111470-100G
A111470-5G
103797-840EA
9
USD
103797-840
103823-660
76869-728
77310-462
4-Chloro-N-methylpicolinamide hydrochloride 97%
4-Chloro-N-methylpicolinamide hydrochloride
Precaución: for Research Use Only
Formula:
C₇H₈Cl₂N₂O MW: 207.06 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD13368182 CAS Number: 882167-77-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...