4-Chloropyridine-3-carboxaldehyde 97%

Supplier: Ambeed
Warning

A129251-10G A129251-250MG A129251-1G A129251-5G A129251-25G
103801-246EA 9 USD
103801-246 77149-534 77149-536 77313-618 77313-620 77685-122
4-Chloropyridine-3-carboxaldehyde 97%
4-Chloropyridine-3-carboxaldehyde

Precaución: for Research Use Only

Formula: C₆H₄ClNO
Boiling Pt: 228.8 °C (760 mm Hg)
Storage Temperature: Freezer
MDL Number: MFCD04112523
CAS Number: 114077-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR