4-Chlorooxindole 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloroindolin-2-one

A167416-100MG A167416-250MG A167416-1G A167416-5G
103806-182EA 31 USD
103806-182 103844-444 77011-468 77011-470 77081-458 77081-460
4-Chlorooxindole 98%
4-Chlorooxindole

Precaución: for Research Use Only

Formula: C₈H₆ClNO
MW: 167.5900 g/mol
Boiling Pt: 340.3 °C (760 mm Hg)
Melting Pt: 206…208 °C
Flash Pt: 159.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03787567
CAS Number: 20870-77-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR