4-Chloropyrocatechol 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-1,2-dihydroxybenzene, 4-Chlorobenzene-1,2-diol, 4-Chlorocatechol

A445861-1G A445861-25G A445861-5G A445861-100G
103827-732EA 25 USD
103827-732 103838-796 76842-588 76842-590 76842-592 77291-646
4-Chloropyrocatechol 98%
4-Chloropyrocatechol

Precaución: for Research Use Only

Formula: C₆H₅ClO₂
MW: 144.56 g/mol
Boiling Pt: 267.1 °C (760 mm Hg)
Melting Pt: 90…94 °C (760 mm Hg)
Flash Pt: 115.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00059614
CAS Number: 2138-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR