4-Chlorobutyraldehyde diethyl acetal 98%

Supplier: Ambeed
Danger

Synonyms: 4-Chloro-1,1-diethoxybutane, 4-Chlorobutanal diethyl acetal

A307594-5G A307594-25G A307594-10G
103835-686EA 15 USD
103835-686 76838-432 77305-958 77672-556
4-Chlorobutyraldehyde diethyl acetal 98%
4-Chlorobutyraldehyde diethyl acetal

Precaución: for Research Use Only

Formula: C₈H₁₇ClO₂
MW: 180.67 g/mol
Boiling Pt: 208.6 °C (760 mm Hg)
Flash Pt: 51.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00671476
CAS Number: 6139-83-9
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR