4-Chlorobenzo[d]thiazole 95%

Supplier: Ambeed
Warning

A185214-100MG A185214-10G A185214-250MG A185214-1G A185214-5G
103815-830EA 22 USD
103815-830 103846-398 76784-710 76784-712 76784-714 77084-032 77084-034
4-Chlorobenzo[d]thiazole 95%
4-Chlorobenzo[d]thiazole

Precaución: for Research Use Only

Formula: C₇H₄ClNS
Boiling Pt: 267.8 °C (760 mm Hg)
Flash Pt: 115.8 °C
Storage Temperature: Ambient
MDL Number: MFCD11040158
CAS Number: 3048-45-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR