4-Chlorobenzo[b]thiophene-2-carboxylic acid 90%

Supplier: Ambeed
Warning

A412669-1G A412669-5G A412669-10G
103827-250EA 7 USD
103827-250 103838-296 76841-842 76841-844 76990-350
4-Chlorobenzo[b]thiophene-2-carboxylic acid 90%
4-Chlorobenzo[b]thiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₅ClO₂S
Boiling Pt: 408.6 °C (760 mm Hg)
Melting Pt: 233 °C
Flash Pt: 200.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06655155
CAS Number: 23967-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR