4-Chlorobenzamidine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 4-Chlorophenylamidine hydrochloride, 4-Chlorobenzene-1-carboximidamide hydrochloride

A371553-1G A371553-5G A371553-100G A371553-10G
103826-484EA 6 USD
103826-484 103837-502 76840-752 76840-754 77142-480 77142-482
4-Chlorobenzamidine hydrochloride 98%
4-Chlorobenzamidine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₈Cl₂N₂
MW: 191.06 g/mol
Melting Pt: 244 °C
Flash Pt: 99.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00126401
CAS Number: 14401-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR