4-Chloro-7-methylquinazoline 95%

Supplier: Ambeed
Warning

A166959-250MG A166959-1G A166959-100MG
103806-114EA 66 USD
103806-114 103844-376 76813-058 77300-862 77671-356
4-Chloro-7-methylquinazoline 95%
4-Chloro-7-methylquinazoline

Precaución: for Research Use Only

Formula: C₉H₇ClN₂
MW: 178.62 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10697149
CAS Number: 90272-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR