4-Chloro-7-methoxyquinoline 98%
Supplier: Ambeed
|
Warning
|
A170311-10G
A170311-250MG
A170311-1G
A170311-25G
A170311-5G
103806-468EA
20
USD
103806-468
103844-742
76799-354
76799-356
77081-852
77081-854
77081-856
4-Chloro-7-methoxyquinoline 98%
4-Chloro-7-methoxyquinoline
Precaución: for Research Use Only
Formula:
C₁₀H₈ClNO MW: 193.6300 g/mol Boiling Pt: 299.8 °C (760 mm Hg) Melting Pt: 75…77 °C Flash Pt: 135.163 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD06659036 CAS Number: 68500-37-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...