4-Chloro-7-hydroxy-6-methoxyquinoline-3-carbonitrile 97%
Supplier: Ambeed
|
Warning
|
A157791-10G
A157791-250MG
A157791-1G
A157791-5G
103805-044EA
21
USD
103805-044
103843-248
76871-916
76871-918
77319-084
77319-086
4-Chloro-7-hydroxy-6-methoxyquinoline-3-carbonitrile 97%
4-Chloro-7-hydroxy-6-methoxyquinoline-3-carbonitrile
Precaución: for Research Use Only
Formula:
C₁₁H₇ClN₂O₂ Boiling Pt: 439 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD09833973 CAS Number: 263149-10-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...