4-Chloro-7H-pyrrolo[2,3-d]pyrimidine-5-carboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A103313-100MG
A103313-250MG
A103313-25G
A103313-5G
103796-528EA
83
USD
103796-528
103822-232
76780-620
76780-622
76780-624
77309-104
4-Chloro-7H-pyrrolo[2,3-d]pyrimidine-5-carboxylic acid 98%
4-Chloro-7H-pyrrolo[2,3-d]pyrimidine-5-carboxylic acid
Precaución: for Research Use Only
Formula:
C₇H₄ClN₃O₂ MW: 197.5800 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11518908 CAS Number: 186519-92-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...