4-Chloro-7-fluoroquinoline 95%

Supplier: Ambeed
Warning

A101470-250MG A101470-1G A101470-5G
103795-988EA 68 USD
103795-988 103821-676 77093-510 77297-114 77297-116
4-Chloro-7-fluoroquinoline 95%
4-Chloro-7-fluoroquinoline

Precaución: for Research Use Only

Formula: C₉H₅ClFN
MW: 181.5941 g/mol
Boiling Pt: 259.9±20.0 °C (760 mm Hg)
Flash Pt: 111 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00153094
CAS Number: 391-82-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR