4-Chloro-7-fluoroquinazoline 95%

Supplier: Ambeed
Warning

A182189-1G A182189-250MG A182189-5G
103807-692EA 43 USD
103807-692 103846-062 77083-602 77287-280 77301-774
4-Chloro-7-fluoroquinazoline 95%
4-Chloro-7-fluoroquinazoline

Precaución: for Research Use Only

Formula: C₈H₄ClFN₂
MW: 182.5800 g/mol
Boiling Pt: 284.881 °C (760 mm Hg)
Flash Pt: 126.092 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08236729
CAS Number: 16499-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR