4-Chloro-7-(trifluoromethyl)quinazoline 95%

Supplier: Ambeed
Warning

A284414-100MG A284414-5G A284414-250MG
103813-638EA 30 USD
103813-638 103834-770 76787-438 77305-456 77668-264
4-Chloro-7-(trifluoromethyl)quinazoline 95%
4-Chloro-7-(trifluoromethyl)quinazoline

Precaución: for Research Use Only

Formula: C₉H₄ClF₃N₂
Boiling Pt: 285.8 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09837196
CAS Number: 16499-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR