4-Chloro-7-(3-chloropropoxy)-6-methoxyquinoline-3-carbonitrile 95%

Supplier: Ambeed
Warning

A228996-100MG A228996-1G
103819-436EA 52 USD
103819-436 76987-270 77089-154
4-Chloro-7-(3-chloropropoxy)-6-methoxyquinoline-3-carbonitrile 95%
4-Chloro-7-(3-chloropropoxy)-6-methoxyquinoline-3-carbonitrile

Precaución: for Research Use Only

Formula: C₁₄H₁₂Cl₂N₂O₂
MW: 311.16 g/mol
Boiling Pt: 478.1 °C (760 mm Hg)
Flash Pt: 243 °C
Storage Temperature: Ambient
MDL Number: MFCD09833974
CAS Number: 214470-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR